missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAp63 Antibody (TAp63-4.1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11703AF488
This item is not returnable.
View return policy
Description
TAp63 Monoclonal antibody specifically detects TAp63 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| TAp63 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| p63 Transactivation Domain Isoform, Transactivation p63, Transactivation Tumor protein 63 | |
| Recombinant human p63 | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| TAp63-4.1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 8626 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction