missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tapasin Related Protein Antibody (OTI4H5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74443H
This item is not returnable.
View return policy
Description
Tapasin Related Protein Monoclonal specifically detects Tapasin Related Protein in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Tapasin Related Protein | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TAPBPL | |
| Full length recombinant protein of human Tapasin Related Protein (NP_060479) produced in HEK293T cells. | |
| 0.1 mL | |
| 55080.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4H5 | |
| Western Blot, Immunohistochemistry | |
| FLJ10143, TAP binding protein-like, Tapasin-like, TAPASIN-R, tapasin-related protein, TAP-binding protein-like, TAP-binding protein-related protein, TAPBP-RTAP binding protein related, TAPBPRtapasin-like | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction