missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAPBPL Monoclonal Antibody (OTI2A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505552S
This item is not returnable.
View return policy
Description
TAPBPL Monoclonal antibody specifically detects TAPBPL in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| TAPBPL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BX59 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55080 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A4 | |
| Unconjugated | |
| TAPBPL | |
| BC017613; TAP binding protein like; TAP binding protein related; TAP binding protein-like; tapasin-like; TAPASIN-R; tapasin-related protein; tapasin-related protein-like; TAP-binding protein-like; TAP-binding protein-related protein; Tapbpl; Tapbplr; TAPBPL-R; TAPBPR; TAPBP-R | |
| Full length human recombinant protein of human TAPBPL produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction