Learn More
Micronova™ Tape Controlled Environment
Shop All Micronova Products
Click to view available options
Color:
Black
Unit Size:
Each
Description
polyethylene, dark green 25mm x 33m Premium
Specifications
Specifications
| Color | Black |
| Format | Roll |
| Material | Polyethylene |
| Sterility | Non-sterile |
| For Use With (Application) | Cleanroom use - color-coding, sealing, labelling and masking, or for pressure-sensitive applications |
| Length (Metric) | 33 m |
| Width (Metric) | 2.5 cm |