missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TARBP1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76400C
This item is not returnable.
View return policy
Description
TARBP1 Polyclonal antibody specifically detects TARBP1 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| TARBP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| DyLight 650 | |
| EC 2.1.1.-, FLJ30482, TAR (HIV-1) RNA binding protein 1, TAR RNA loop binding protein, TAR RNA-binding protein 1, TAR RNA-binding protein of 185 kDa, tardbp1, TRM3probable methyltransferase TARBP1, TRP-185TAR (HIV) RNA binding protein 1, TRP185TAR (HIV) RNA-binding protein 1 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the center of human TRBP1. The immunogen is located within amino acids 1310 - 1360 of TRBP1. Amino Acid Squence: HGAGNAKKNWQRIQEHF | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 6894 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction