missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TARBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 572.00
Specifications
| Antigen | TARBP1 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18483071
|
Novus Biologicals
NBP1-86902 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18413761
|
Novus Biologicals
NBP1-86902-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TARBP1 Polyclonal antibody specifically detects TARBP1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| TARBP1 | |
| Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Infections (Virus Bacteria and Parasites) | |
| PBS (pH 7.2) and 40% Glycerol | |
| 6894 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.1.1.-, FLJ30482, TAR (HIV-1) RNA binding protein 1, TAR RNA loop binding protein, TAR RNA-binding protein 1, TAR RNA-binding protein of 185 kDa, tardbp1, TRM3probable methyltransferase TARBP1, TRP-185TAR (HIV) RNA binding protein 1, TRP185TAR (HIV) RNA-binding protein 1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: MNELNSVSDLDRCHLYLMVLTELINLHLKVGWKRGNPIWRVISLLKNASIQHLQEMDSGQEPTVGSQIQRVVSMAALAMVCEAIDQKPELQLDSLHAG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title