missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TARBP2 Monoclonal Antibody (OTI7C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808481
This item is not returnable.
View return policy
Description
TRBP is an RNA associated nuclear protein. It contains 3 DRBM (double stranded RNA-binding) domains and plays an important role by binding to HIV-1 TAR RNA located in the long terminal repeat (LTR).TRBP acts synergistic with viral TAT protein in HIV-1 expression activation. It binds preferentially to TAR and other double-stranded RNAs with G+C-rich characteristics. It is important in regulation of transcription from RNA polymerase II promoter andis widely expressed in most of the tissues.Specifications
| TARBP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15633 | |
| TARBP2 | |
| Full length human recombit protein of human TARBP2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7C1 | |
| Unconjugated | |
| TARBP2 | |
| LOQS, TRBP, TRBP1, TRBP2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6895 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction