missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TARSH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 624.00
Specifications
| Antigen | TARSH |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636485
|
Novus Biologicals
NBP2-48811-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600736
|
Novus Biologicals
NBP2-48811 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TARSH Polyclonal antibody specifically detects TARSH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TARSH | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ABI family, member 3 (NESH) binding protein, ABI gene family member 3-binding protein, DKFZP586L2024, FLJ41743, Nesh-binding protein, NeshBP, NESHBPABI gene family, member 3 (NESH) binding protein, target of Nesh-SH3, Tarsh, TARSHFLJ41754 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VEGGIWSKIFNHKTVVGSKKVNGKIQSTYDQDHTVPAYVPRKLIPITIIKQVIQNVTHKDSAKSPEKAPLGGVILVHLIIPGLNETTVKLPASLMFEISDAL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 25890 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title