Learn More
TAS-103 dihydrochloride, MedChemExpress
Product Code. 16436316
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16436316 100 mg 100mg
16425848 10 mg 10mg
16411997 50 mg 50mg
16408507 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16436316 Supplier Medchem Express Supplier No. HY13758A
MedChemExpress TAS-103 dihydrochloride is a dual inhibitor of DNA topoisomerase I/II, used for cancer research.
TAS-103 dihydrochloride is a dual inhibitor of DNA topoisomerase I/II, used for cancer research.
Specifications
Chemical Name or Material TAS-103 dihydrochloride
CAS 174634-09-4
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C20H21Cl2N3O2
Quantity 100 mg
Synonym BMS-247615 dihydrochloride
Solubility Information H2O : 50 mg/mL (123.06 mM; Need ultrasonic)
SMILES O=C(C1=C2C3=CC=C(O)C=C3N=C1NCCN(C)C)C4=C2C=CC=C4.Cl.Cl
Molecular Weight (g/mol) 406.31
Formula Weight 406.31
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only