missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS1R3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS5060
This item is not returnable.
View return policy
Description
TAS1R3 Polyclonal antibody specifically detects TAS1R3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TAS1R3 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| T1R3Sweet taste receptor T1R3, taste receptor type 1 member 3, taste receptor, type 1, member 3, TR3 | |
| Synthetic 19 amino acid peptide from N-terminal extracellular domain of human TAS1R3. | |
| 0.05 mL | |
| GPCR | |
| 83756 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 10 - 20 μg/mL | |
| Q7RTX0 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction