missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAS2R30 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567773
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Predicted to react with Mouse and Rat samples.
Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. [UniProt].
Especificaciones
| TAS2R30 | |
| Polyclonal | |
| Unconjugated | |
| TAS2R30 | |
| T2R30; T2R44; T2R47; T2R53; TAS2R30; TAS2R44; TAS2R47; taste 2 receptor member 30; taste receptor T2R47; taste receptor type 2 member 30; Taste receptor type 2 member 47; taste receptor, type 2, member 30; taste receptor, type 2, member 47; type 2 taste receptor member 30; type 2 taste receptor member 47 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 259293 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P59541 | |
| TAS2R30 | |
| Synthesized peptide derived from human TAS2R30. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido