missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAS2R46 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16624205
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16624205 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16624205 Supplier Invitrogen™ Supplier No. PA599956

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAS2R46.

Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. Expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Most taste cells may be activated by a limited number of bitter compounds; individual taste cells can discriminate among bitter stimuli.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R46
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAS2R46
Gene Accession No. P59540
Gene Alias MGC134865; T2R46; T2R54; TAS2R46; TAS2R47; taste 2 receptor member 46; taste receptor type 2 member 46; Taste receptor type 2 member 54; taste receptor, type 2, member 46
Gene Symbols TAS2R46
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAS2R46(Accession P59540), corresponding to amino acid residues F195-I245.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259292
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.