Learn More
Tashiro's indicator solution, In ethanol, Honeywell Fluka™
Product Code. 15658390 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
30 mL
250 mL
1 L
1.5 L
6 x 250 mL
6 x 1 L
Unit Size:
1 L
1.5 L
250mL
30mL
Pack of 6
Product Code. 15658390
Brand: Honeywell-Fluka 3608330ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
In ethanol
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tashiro’s indicator solution
Quantity 30 mL
Flash Point 12°C (53.6°F)