Learn More
TASP1 Mouse anti-Human, Clone: OTI1H6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502257
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes an endopeptidase that cleaves specific substrates following aspartate residues. The encoded protein undergoes posttranslational autoproteolytic processing to generate alpha and beta subunits, which reassemble into the active alpha2-beta2 heterotetramer. It is required to cleave MLL, a protein required for the maintece of HOX gene expression, and TFIIA, a basal transcription factor. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Specifications
| TASP1 | |
| Monoclonal | |
| Unconjugated | |
| TASP1 | |
| C20orf13, dJ585I14.2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55617 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1H6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9H6P5 | |
| TASP1 | |
| Full length human recombit protein of human TASP1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.