missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAT226 Antibody (Genentech patent anti-TAT226), Janelia Fluor™ 549, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27958JF549
This item is not returnable.
View return policy
Description
TAT226 Monoclonal antibody specifically detects TAT226 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| TAT226 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Human | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Genentech patent anti-TAT226 | |
| 50mM Sodium Borate | |
| TAT226 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction