Learn More
TAT226 Antibody (Genentech patent anti-TAT226), mFluor Violet 610 SE, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-27958MFV610
Product Code. 30116423
  • € 475.00 / 0.10mL
Description
TAT226 Monoclonal antibody specifically detects TAT226 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TAT226
Monoclonal
mFluor Violet 610 SE
Human
Protein A purified
RUO
Human
Purified
Flow Cytometry, ELISA, Functional Assay
Genentech patent anti-TAT226
50mM Sodium Borate
TAT226
0.1 mL
Primary
Store at 4°C in the dark.