Learn More
TATA binding protein TBP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95183-0.1ml
This item is not returnable.
View return policy
Description
TATA binding protein TBP Polyclonal antibody specifically detects TATA binding protein TBP in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| TATA binding protein TBP | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| GTF2D, GTF2D1, HDL4, MGC126054, MGC126055, SCA17, TATA box binding protein, TATA sequence-binding protein, TATA-binding factor, TATA-box binding protein N-terminal domain, TATA-box factor, TATA-box-binding protein, TF2D, TFIIDMGC117320, Transcription initiation factor TFIID TBP subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-319 of human TBP (NP_001165556.1). LKTIALRARNAEYNPKRFAAVIMRIREPRTTALIFSSGKMVCTGAKSEEQSRLAARKYARVVQKLGFPAKFLDFKIQNMVGSCDVKFPIRLEGLVLTHQQFSSYEPELFPGLIYRMIKPRIVLLIFVSGKVVLTGAKVRAEIYEAFENIYPILKGFRKTT | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 6908 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |