missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TATA binding protein TBP Antibody (SR1605), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 472.00
Specifications
| Antigen | TATA binding protein TBP |
|---|---|
| Clone | SR1605 |
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696714
|
Novus Biologicals
NBP3-21665-100ul |
100 μg |
€ 472.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665164
|
Novus Biologicals
NBP3-21665-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TATA binding protein TBP Monoclonal antibody specifically detects TATA binding protein TBP in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TATA binding protein TBP | |
| Western Blot 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| GTF2D, GTF2D1, HDL4, MGC126054, MGC126055, SCA17, TATA box binding protein, TATA sequence-binding protein, TATA-binding factor, TATA-box binding protein N-terminal domain, TATA-box factor, TATA-box-binding protein, TF2D, TFIIDMGC117320, Transcription initiation factor TFIID TBP subunit | |
| A synthesized peptide derived from human TATA binding protein TBP (Uniprot #: P20226) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1605 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6908 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title