missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TATDN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584140
This item is not returnable.
View return policy
Description
Immunogen sequence: MSRFKFIDIG INLTDPMFRG IYRGVQKHQD DLQDVIGRAV EIGVKKFMIT GGNLQDSKDA LHLAQTNGMF Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 89%.
TATDN1 is a protein coding gene. Putative deoxyribonuclease.
Specifications
| TATDN1 | |
| Polyclonal | |
| Unconjugated | |
| TATDN1 | |
| 2310079P03Rik; CDA11; hepatocarcinoma high expression protein; LOC690068; Putative deoxyribonuclease TATDN1; similar to TatD DNase domain containing 1; TatD DNase domain containing 1; Tatdn1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83940 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P1N9 | |
| TATDN1 | |
| Recombinant protein corresponding to Human TATDN1. Recombinant protein control fragment (Product #RP-104959). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction