missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau Antibody (2E9), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25162UV
This item is not returnable.
View return policy
Description
Tau Monoclonal specifically detects Tau in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Tau | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MAPT | |
| Recombinant full length 441 amino acid human tau. Epitope located in the peptide KDRVQSKIGSLDNITHVPGG, amino acids 347-366. | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Chaperone Mediated Autophagy (CMA), Cytokine Research, Cytoskeleton Markers, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers | |
| 4137 | |
| Human, Mouse, Rat, Pig, Bovine, Equine | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 2000000000 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to bind to all Tau isoforms | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction