missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau Oligomeric Complex Antibody (TOC1), DyLight 755 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20163IR
This item is not returnable.
View return policy
Description
Tau Oligomeric Complex Monoclonal antibody specifically detects Tau Oligomeric Complex in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| Tau Oligomeric Complex | |
| Monoclonal | |
| DyLight 755 | |
| Tau Oligomeric Complex 1, TOC1, TOC-1 | |
| The original antibody was generated by immunizing a mouse with electro eluted cross-linked Tau dimers. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| TOC1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction