missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau, p Ser202 Antibody (S01-5A7), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 645.00
Specifications
| Antigen | Tau |
|---|---|
| Clone | S01-5A7 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682225
|
Novus Biologicals
NBP3-20058-100ul |
100 μg |
€ 645.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644005
|
Novus Biologicals
NBP3-20058-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tau Monoclonal antibody specifically detects Tau in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Tau | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
| A synthetic phosphopeptide corresponding to residues surrounding Ser202 of human Tau (Uniprot # P10636) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S01-5A7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 4137 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title