missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau, p Ser404 Antibody (11H10), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | Tau |
|---|---|
| Clone | 11H10 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30025479
|
Novus Biologicals
NBP3-26408-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30032973
|
Novus Biologicals
NBP3-26408-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tau Monoclonal antibody specifically detects Tau in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| Tau | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
| A synthesized peptide derived from Human Tau [p Ser404] [UniProt P10636] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 11H10 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 4137 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title