Learn More
TAU Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-10274-100UL
 View more versions of this product
Product Code. 18055565
  • € 450.00 / 100µL
Description
The TAU antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TAU. This antibody recognizes human, mouse, rat antigen. The TAU antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
TAU
1000 μg/mL
DDPAC, FLJ31424, FTDP 17, MAPT, MAPTL, MSTD, MTBT1, MTBT2, Neurofibrillary tangle protein, Paired helical filament tau, PHF tau, PPND, TAU
100 μL
4137
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa