Learn More
TAU Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-10274-100UL
Description
The TAU antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TAU. This antibody recognizes human, mouse, rat antigen. The TAU antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| TAU | |
| 1000 μg/mL | |
| DDPAC, FLJ31424, FTDP 17, MAPT, MAPTL, MSTD, MTBT1, MTBT2, Neurofibrillary tangle protein, Paired helical filament tau, PHF tau, PPND, TAU | |
| 100 μL | |
| 4137 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |