missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAX1BP3 Monoclonal Antibody (1H5)

TAX1BP3 Monoclonal Antibody (1H5)
Brand: Invitrogen MA521820
This item is not returnable.
View return policy
Description
Specifications
| TAX1BP3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O14907 | |
| TAX1BP3 | |
| TAX1BP3 (AAH23980, 1 a.a. ~ 124 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1H5 | |
| Unconjugated | |
| TAX1BP3 | |
| 1300011C24Rik; C20orf100; EMBL:AAH97976.1}; GCX1; glutaminase-interacting protein 3; Tax interaction protein 1; Tax1 (human T cell leukemia virus type I) binding protein 3; Tax1 (human T-cell leukemia virus type I) binding protein 3; Tax1 binding protein 3; tax1-binding protein 3; TAX1BP3; tax1bp3 {ECO:0000312; tax-interacting protein 1; TIP1; TIP-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 30851 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction