missing translation for 'onlineSavingsMsg'
Learn More
Learn More
taxilin gamma Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22330AF532
This item is not returnable.
View return policy
Description
taxilin gamma Polyclonal antibody specifically detects taxilin gamma in Human samples. It is validated for Western Blot
Specifications
| taxilin gamma | |
| Polyclonal | |
| Western Blot | |
| ELRG, factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription, FIAT, lipopolysaccharide-specific response protein 5, taxilin gamma | |
| The immunogen this antibody was made to, maps to a region between residue 475 to 525 of human Taxilin Gamma using the numbering given in entry NP_060830.2 (GeneID 55787). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55787 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction