missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Taxol Janelia Fluor 646

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Taxol Janelia Fluor® 646 is a red fluorescent taxol derivative for direct imaging of the microtubule cytoskeleton. Taxol Janelia Fluor® 646 is fluorogenic: fluoresces only once bound to microtubules, enabling hassle-free no-wash experiments. Excitation maximum = 655 nm; emission maximum = 671 nm.
For more information on how Taxol-Janelia Fluor™ 646 may be used, see our protocol .
Specifications
Specifications
| Chemical Name or Material | 2-(3-(Azetidin-1-ium-1-ylidene)-7-(azetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-(((2 S)-1-(((2 aR,4 S,4 AS,6 R,9 S,11 S,12 S,12 bS)-6,12 b-diacetoxy-9-(((2 R,3 S)-3-benzamido-2-hydroxy-3-phenylpropanoyl)oxy)-12-(benzoyloxy)-11-hydroxy-4 A,8,13,13- |
| Quantity | 1 mg |
| Target | Microtubule Probes |
| Molecular Formula | C79H82N4O18Si |
| Formula Weight | 1403.62 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction