missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAZ Polyclonal Antibody
GREENER_CHOICE

Product Code. 30019770
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019770 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30019770 Supplier Invitrogen™ Supplier No. PA5148202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TAZ Polyclonal Antibody for IHC (P)

This gene encodes a protein that is expressed at high levels in cardiac and skeletal muscle. Mutations in this gene have been associated with a number of clinical disorders including Barth syndrome, dilated cardiomyopathy, hypertrophic DCM, endocardial fibroelastosis, and left ventricular noncompaction. Multiple transcript variants encoding different isoforms have been described. A long form and a short form of each of these isoforms is produced; the short form lacks a hydrophobic leader sequence and may exist as a cytoplasmic protein rather than being membrane-bound. Other alternatively spliced transcripts have been described but the full-length nature of all these transcripts is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAZ
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TAZ
Gene Accession No. Q16635
Gene Alias 2310058J06Rik; 2610021I22Rik; 5031411C02Rik; 9130012G04Rik; AW107266; AW552613; Barth syndrome); BTHS; C78399; cardiomyopathy, dilated 3A (X-linked); CMD3A; EFE; EFE2; endocardial fibroelastosis 2; FLJ27390; G4.5; HGNC:11577; LVNCX; OTTHUMP00000061673; protein G4.5; Tafazzin; tafazzin (cardiomyopathy, dilated 3A (X-linked); tafazzin (cardiomyopathy, dilated 3A (X-linked); endocardial fibroelastosis 2; Barth syndrome); Taz; Taz1; transcriptional coactivator with PDZ binding motif; transcriptional co-activator with PDZ-binding motif; transcriptional coactivator with PDZ-binding motif; transcriptional co-activator with PDZ-binding motif (TA)Z; transcriptional co-activator with PDZ-binding motif (TAZ); WW domain containing transcription regulator 1; WW domain-containing transcription regulator protein 1; Wwtr1; XAP-2; XX-FW83563B9.3
Gene Symbols TAZ
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the Human TAZ.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6901
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.