missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAZ/WWTR1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-220MFV450
This item is not returnable.
View return policy
Description
TAZ/WWTR1 Polyclonal antibody specifically detects TAZ/WWTR1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TAZ/WWTR1 | |
| Polyclonal | |
| Western Blot | |
| FLJ27004, FLJ45718, TAZDKFZp586I1419, transcriptional co-activator with PDZ-binding motif, Transcriptional coactivator with PDZ-binding motif, WW domain containing transcription regulator 1, WW domain-containing transcription regulator protein 1 | |
| Synthetic peptide representing residues 386-400 of the human TAZ protein. | |
| 0.1 mL | |
| Chromatin Research, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators | |
| 25937 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction