missing translation for 'onlineSavingsMsg'
Learn More

TAZ/WWTR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

€ 271.00 - € 572.00

Specifications

Antigen TAZ/WWTR1
Dilution Western Blot 0.4 ug/ml, Simple Western 1:30, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18449250
View Documents
Novus Biologicals
NBP1-85067-25ul
25ul
€ 271.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18739223
View Documents
Novus Biologicals
NBP1-85067
€ 572.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TAZ/WWTR1 Polyclonal specifically detects TAZ/WWTR1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

TAZ/WWTR1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Unconjugated
RUO
Human, Mouse
Q9GZV5
25937
This antibody was developed against Recombinant Protein corresponding to amino acids:MNPKPSSWRKKILPESFFKEPDSGSHSRQSSTDSSGGHPGPRLAGGAQHVRSHSSPASLQLGTGAGAAGSPAQQHAHLRQQSYDVTDELPLPPGWEMTFTATGQRYFLNHIEKITTWQDPRKAMNQPLNHMNLHPAVSST
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 0.4 ug/ml, Simple Western 1:30, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated
Polyclonal
Rabbit
Chromatin Research, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
FLJ27004, FLJ45718, TAZDKFZp586I1419, transcriptional co-activator with PDZ-binding motif, Transcriptional coactivator with PDZ-binding motif, WW domain containing transcription regulator 1, WW domain-containing transcription regulator protein 1
WWTR1
IgG
Affinity Purified
Specificity of human TAZ/WWTR1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.