Learn More
Tazobactam, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Tazobactam |
| CAS | 89786-04-9 |
| Analyte or Component Names | Tazobactam |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 N4 O5 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3-methyl-7-oxo-3-(1H-1,2,3-triazol-1-ylmethyl)-, 4,4-dioxide, (2S,3S,5R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3-methyl-7-oxo-3-(1H-1,2,3-triazol-1-ylmethyl)-, 4,4-dioxide, [2S-(2α,3β,5α)]-, 1H-1,2,3-Triazole, 4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid deriv., CL 298741, YTR 830H |
| InChI Formula | InChI=1S/C10H12N4O5S/c1-10(5-13-3-2-11-12-13)8(9(16)17)14-6(15)4-7(14)20(10,18)19/h2-3,7-8H,4-5H2,1H3,(H,16,17)/t7-,8+,10+/m1/s1 |
| Solution Type | Neat |
| SMILES | C([C@@]1(C)[C@H](C(O)=O)N2[C@](S1(=O)=O)(CC2=O)[H])N3C=CN=N3 |
| Show More |