missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21442MFV610
This item is not returnable.
View return policy
Description
TBC1D1 Polyclonal antibody specifically detects TBC1D1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TBC1D1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| KIAA1108TBC, TBC1, TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1, TBC1 domain family member 1 | |
| Produced in rabbits immunized with E. coli-derived Human TBC1D1 fragment | |
| 0.1 mL | |
| Signal Transduction | |
| 23216 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction