missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TBC1D1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29370
This item is not returnable.
View return policy
Description
TBC1D1 Polyclonal antibody specifically detects TBC1D1 in Human samples. It is validated for Immunoprecipitation
Specifications
| TBC1D1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| KIAA1108TBC, TBC1, TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1, TBC1 domain family member 1 | |
| Between 500 and 550 | |
| 100 μg | |
| Signal Transduction | |
| 23216 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction