missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D10A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00
Specifications
| Antigen | TBC1D10A |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
TBC1D10A Polyclonal specifically detects TBC1D10A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| TBC1D10A | |
| Polyclonal | |
| Rabbit | |
| Human | |
| EBP50-PDX interactor of 64 kDa, EPI64, rab27A-GAP-alpha, TBC1 domain family member 10A, TBC1 domain family, member 10A | |
| TBC1D10A | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 83874 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:('KPKPPKQAQKEQRKQMKGRGQLEKPPAPNQAMVVAAAGDACPPQHVPPKDSAPKDSAPQDLAPQVSAHHRSQESLTSQESEDTYL',) | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title