missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D13 Antibody (OTI2C5), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74450AF750
This item is not returnable.
View return policy
Description
TBC1D13 Monoclonal specifically detects TBC1D13 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TBC1D13 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TBC1D13 | |
| Full length recombinant protein of human TBC1D13 (NP_060671) produced in HEK293T cells. | |
| Signal Transduction | |
| 54662 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2C5 | |
| Western Blot, Immunohistochemistry | |
| FLJ10743, TBC1 domain family member 13, TBC1 domain family, member 13 | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction