missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D13 Monoclonal Antibody (OTI2C5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526207
This item is not returnable.
View return policy
Description
TBC1D13 Monoclonal Antibody for Western Blot, IHC (P)
TBC1D13 may act as a GTPase-activating protein for Rab family protein(s).
Specifications
| TBC1D13 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NVG8 | |
| TBC1D13 | |
| Full length human recombinant protein of TBC1D13 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C5 | |
| Unconjugated | |
| TBC1D13 | |
| 2600014A06Rik; BC025586; TBC1 domain family member 13; TBC1 domain family, member 13; TBC1D13 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54662 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction