Learn More
Invitrogen™ TBC1D13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114139
Description
Immunogen sequence: FLLLVCCAML MLIREQLLEG DFTVNMRLLQ DYPITDVCQI LQKAKELQDS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
TBC1D13 may act as a GTPase-activating protein for Rab family protein(s).
Specifications
| TBC1D13 | |
| Polyclonal | |
| Unconjugated | |
| TBC1D13 | |
| 2600014A06Rik; BC025586; TBC1 domain family member 13; TBC1 domain family, member 13; TBC1D13 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54662 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NVG8 | |
| TBC1D13 | |
| Synthetic peptide directed towards the middle region of human TBC1D13. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.