Learn More
TBC1D15 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-36552AF700
Product Code. 18727850
  • € 633.00 / 0.10mL
Description
TBC1D15 Polyclonal antibody specifically detects TBC1D15 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TBC1D15
Western Blot
Rabbit
The immunogen recognized by the antibody maps to a region between residue 1 to 50 of human TBC1 Domain Family, Member 15 using the numbering given in entry NP_073608.4.