missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D15 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36552H
This item is not returnable.
View return policy
Description
TBC1D15 Polyclonal antibody specifically detects TBC1D15 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TBC1D15 | |
| Polyclonal | |
| Western Blot | |
| domain family member 15, TBC1 domain family, member 15 | |
| The immunogen recognized by the antibody maps to a region between residue 1 to 50 of human TBC1 Domain Family, Member 15 using the numbering given in entry NP_073608.4. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 64786 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction