missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D15 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36553JF549
This item is not returnable.
View return policy
Description
TBC1D15 Polyclonal antibody specifically detects TBC1D15 in Human samples. It is validated for Western Blot
Specifications
| TBC1D15 | |
| Polyclonal | |
| Western Blot | |
| Immunogen affinity purified | |
| Store at 4°C in the dark. |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| RUO | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction