missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566800
This item is not returnable.
View return policy
Description
Immunogen sequence: FFGGIDVSIRG EVWPFLLRYY SHESTSEERE ALRLQKRKEY SEIQQKRLSM TPEEHRAFWR NVQFTVDKDV VRTDRNNQFF RGEDNPNVES MRRILLNYAV YNPAV Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
May act as a GTPase-activating protein for Rab family protein(s).
Specifications
| TBC1D16 | |
| Polyclonal | |
| Unconjugated | |
| TBC1D16 | |
| CTD-2529O21.1; TBC1 domain family member 16; TBC1 domain family, member 16; TBC1D16 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 125058 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TBP0 | |
| TBC1D16 | |
| Recombinant Human TBC1D16. Recombinant protein control fragment (Product #RP-105372). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction