missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D21 Antibody (OTI2A6), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74451APCCY7
This item is not returnable.
View return policy
Description
TBC1D21 Monoclonal specifically detects TBC1D21 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| TBC1D21 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| TBC1D21 | |
| Full length recombinant protein of human TBC1D21 (NP_699187) produced in HEK293T cells. | |
| 0.1 mL | |
| 161514.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI2A6 | |
| Flow Cytometry | |
| MGC34741, TBC1 domain family member 21, TBC1 domain family, member 21 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction