missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D21 Monoclonal Antibody (OTI7B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503762
This item is not returnable.
View return policy
Description
TBC1D21 may act as a GTPase-activating protein for Rab family protein(s). May be involved in acrosome formation and cytoskeletal reorganization during spermiogenesis, possibly by regulating RAB3A activity.Specifications
| TBC1D21 | |
| Monoclonal | |
| 0.47 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IYX1 | |
| TBC1D21 | |
| Full length human recombit protein of human TBC1D21 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7B1 | |
| Unconjugated | |
| TBC1D21 | |
| MgcRabGAP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 161514 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction