Learn More
TBC1D21 Mouse anti-Human, Clone: OTI3F1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503742
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TBC1D21 may act as a GTPase-activating protein for Rab family protein(s). May be involved in acrosome formation and cytoskeletal reorganization during spermiogenesis, possibly by regulating RAB3A activity.Specifications
| TBC1D21 | |
| Monoclonal | |
| Unconjugated | |
| TBC1D21 | |
| MgcRabGAP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 161514 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3F1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8IYX1 | |
| TBC1D21 | |
| Full length recombit protein of human TBC1D21 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.