missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D22A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20578
This item is not returnable.
View return policy
Description
TBC1D22A Polyclonal specifically detects TBC1D22A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TBC1D22A | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| HSC79E021, TBC1 domain family member 22A, TBC1 domain family, member 22A | |
| Rabbit | |
| 59 kDa | |
| 0.1 mL | |
| Primary | |
| Species reactivity based on sequence identity: Mouse (85%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q8WUA7 | |
| TBC1D22A | |
| Recombinant protein encompassing a sequence within the center region of human TBC1D22A. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 25771 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction