missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D23 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68409-1-IG-20UL
This item is not returnable.
View return policy
Description
The TBC1D23 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TBC1D23. This antibody recognizes human, mouse, rat, pig antigen. The TBC1D23 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| TBC1D23 | |
| Monoclonal | |
| 1000 μg/mL | |
| DKFZp667G062; FLJ11046; NS4ATP1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 78 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 2A12A7 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 55773 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction