Learn More
TBC1D24 Mouse, anti-Human, Clone: 1C7C4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68566-1-IG-150UL
 View more versions of this product
Product Code. 18045585
  • € 410.00 / 150µL
Description
The TBC1D24 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TBC1D24. This antibody recognizes human, rat, mouse antigen. The TBC1D24 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TBC1D24
1C7C4
KIAA1171, TBC1 domain family member 24, TBC1 domain family, member 24, TBC1D24
150 μL
57465
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
150 kDa