Learn More
TBC1D24 Mouse, anti-Human, Clone: 1C7C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68566-1-IG-150UL
Description
The TBC1D24 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TBC1D24. This antibody recognizes human, rat, mouse antigen. The TBC1D24 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TBC1D24 | |
| 1C7C4 | |
| KIAA1171, TBC1 domain family member 24, TBC1 domain family, member 24, TBC1D24 | |
| 150 μL | |
| 57465 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse | |
| 150 kDa | |
| IgG1 |