missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D28 Antibody (OTI4A9), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74461V
This item is not returnable.
View return policy
Description
TBC1D28 Monoclonal specifically detects TBC1D28 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TBC1D28 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TBC1D28 | |
| Full length recombinant protein of human TBC1D28 (NP_001034486) produced in E.coli. | |
| 0.1 mL | |
| 254272.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4A9 | |
| Western Blot, Immunohistochemistry | |
| FLJ40244, MGC138217, MGC138221, TBC1 domain family member 28, TBC1 domain family, member 28 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction