missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | TBC1D4 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30226573
|
Novus Biologicals
NBP3-35805-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229275
|
Novus Biologicals
NBP3-35805-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBC1D4 Polyclonal antibody specifically detects TBC1D4 in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| TBC1D4 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, B Cell Development and Differentiation Markers, Diabetes Research, Lipid and Metabolism, Phospho Specific, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 9882 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1059-1298 of human TBC1D4 (NP_055647.2).,, Sequence:, SRLLHDYHRDLYNHLEENEISPSLYAAPWFLTLFASQFSLGFVARVFDIIFLQGTEVIFKVALSLLSSQETLIMECESFENIVEFLKNTLPDMNTSEMEKIITQVFEMDISKQLHAYEVEYHVLQDELQESSYSCEDSETLEKLERANSQLKRQNMDLLEKLQVAHTKIQALESNLENLLTRETKMKSLIRTLEQEKMAYQKTVEQLRKLLPADALVNCDLLLRDLNCNPNNKAKIGNKP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title