missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D4 Antibody (OTI5E6), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74462PCP
This item is not returnable.
View return policy
Description
TBC1D4 Monoclonal specifically detects TBC1D4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| TBC1D4 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| TBC1D4 | |
| Full length recombinant protein of human TBC1D4 (NP_0055647) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Flow Cytometry | |
| OTI5E6 | |
| Flow Cytometry | |
| Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4 | |
| Mouse | |
| Affinity Purified | |
| Diabetes Research, Lipid and Metabolism, Phospho Specific | |
| 9882.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction